VWR®, Rack for Cryogenic Vials
Supplier: VWR Collection
VWRI211-0139EA 23.7 SGD
VWRI211-0139
VWR®, Rack for Cryogenic Vials
Racks Cryogenic Vial Racks
PC.
  • Numeric hole index
  • Moulded tabs in the bottom of each well to secure vials in place for easier opening and closing
  • Ribbed grips make handling with gloves easier
Withstands both cryogenic freezing and autoclaving.
Order Now
SPECIFICATIONS
Environmentally Preferable