VWR®, Test Tube Racks, Fold and Snap
Supplier: VWR Collection
VWRI211-0351EA 24.2 SGD
VWRI211-0351 VWRI211-0352 VWRI211-0353 VWRI211-0354 VWRI211-0355 VWRI211-0356 VWRI211-0357 VWRI211-0358 VWRI211-0359 VWRI211-0360 VWRI211-0361 VWRI211-0362
VWR®, Test Tube Racks, Fold and Snap
Racks Tube Racks
PP.
  • Easy to assemble polypropylene racks snap together in seconds
  • Designed to remain stable in water baths
  • Shipped flat
Order Now
SPECIFICATIONS
Environmentally Preferable