VWR® Rota-Rack Duo, Racks, Microcentrifuge Tube
Supplier: VWR Collection
VWRI211-0369EA 115 SGD
VWRI211-0369
VWR® Rota-Rack Duo, Racks, Microcentrifuge Tube
Racks Tube Racks
Work with PCR tubes right next to centrifuge tubes, dilute or take off concentrate using one rack.
  • Rotate half the rack to suit particular requirements
  • Purple module: 48 places for 0,5 ml microtubes or 48 places for 1,5 ml microtubes
  • Blue module: 96 places for PCR tubes or 48 places for 1,5 ml microtubes
  • Plate format is, therefore, compatible with 8-channel multi-pipettes
  • Tapered wells with alphanumeric grid to help identification