VWR® Microban®, Glove Box Holders
Supplier: VWR Collection
VWRI211-0371EA 319 SGD
VWRI211-0371
VWR® Microban®, Glove Box Holders
Glove Dispensers and Glove Box Holders
These wall-mountable glove box holders are made from polystyrene with Microban® additive.
  • Microban® additive prevents growth of moulds and bacteria
  • High impact polystyrene, resistant to harsh cleaning solutions
  • Modular design allows stacking to save space
Suitable for use in labs, cleanrooms, hospitals and microbiological work areas.
Delivery information: Mounting screws included.