VWR®, Racks for 15 to 50 ml Test Tubes
Supplier: VWR Collection
VWRI211-0380EA 154 SGD
VWRI211-0380
VWR®, Racks for 15 to 50 ml Test Tubes
Racks Tube Racks
PP.
  • Holds up to 12 similar size tubes with a diameter from 16 to 30 mm
  • Imprinted references for easy tube identification
  • Not suitable for autoclaving
Quickly change aperture by opening the 'iris' with a turn of the knob, or directly insert a tube into the opening.