VWR®, Test Tubes Racks
Supplier: VWR Collection
89215-768 89215-718
VWRI212-0409EA 11.8 SGD
VWRI212-0409 VWRI212-0422 VWRI212-0421 VWRI212-0420 VWRI212-0419 VWRI212-0418 VWRI212-0417 VWRI212-0416 VWRI212-0415 VWRI212-0414 VWRI212-0413 VWRI212-0412 VWRI212-0411 VWRI212-0425 VWRI212-0407 VWRI212-0393 VWRI212-0406 VWRI212-0405 VWRI212-0404 VWRI212-0403 VWRI212-0402 VWRI212-0401 VWRI212-0400 VWRI212-0399 VWRI212-0398 VWRI212-0397 VWRI212-0396 VWRI212-0382 VWRI212-0394 VWRI212-0410 VWRI212-0392 VWRI212-0391 VWRI212-0390 VWRI212-0389 VWRI212-0388 VWRI212-0387 VWRI212-0386 VWRI212-0385 VWRI212-0384 VWRI212-0383 VWRI212-0395 VWRI212-0423 VWRI212-0430 VWRI212-0429 VWRI212-0428 VWRI212-0427 VWRI212-0426 VWRI212-0408 VWRI212-0424 VWRU89215-768 VWRU89215-718
VWR®, Test Tubes Racks
Racks Tube Racks
Nylon, glass fibre reinforced.
  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Order Now
SPECIFICATIONS
Environmentally Preferable