VWR® Mega, Test Tube Racks
Supplier: VWR Collection
VWRI211-0366EA 124 SGD
VWRI211-0366 VWRI211-0363 VWRI211-0364 VWRI211-0365 VWRI212-0660 VWRI212-0661 VWRI212-0662 VWRI212-0663
VWR® Mega, Test Tube Racks
Racks Tube Racks
High capacity storage for your tubes.
  • Sturdy, material for easy and secure transport of blood collection and other tubes
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack eliminating the need for multiple racks
  • Autoclavable
Fully assembled, polypropylene mega racks are great for high throughput labs requiring large sample storage.
Order Now
SPECIFICATIONS
Environmentally Preferable