VWR®, Test Tubes with Rim
Supplier: VWR Collection
VWRI212-0753EA 19.3 SGD
VWRI212-0753 VWRI212-0754 VWRI212-0755 VWRI212-0756 VWRI212-0757 VWRI212-0758 VWRI212-0759 VWRI212-0760 VWRI212-0761 VWRI212-0762 VWRI212-0763 VWRI212-0764 VWRI212-0765
VWR®, Test Tubes with Rim
Tubes Test Tubes
Test tubes made from borosilicate 5.1 glass, neutral glass.
  • Thin walled
  • Excellent chemical resistance
  • Relatively resistant to rapid temperature changes and local heating
The economical alternative for a wide range of uses.
Certifications: DIN 12395.
Order Now
SPECIFICATIONS
Environmentally Preferable