VWR® Culture Tubes With Screw Thread
Supplier: VWR Collection
VWRI212-0849EA 152 SGD
VWRI212-0849 VWRI212-0850 VWRI212-0851 VWRI212-0852 VWRI212-0853 VWRI212-0854
VWR® Culture Tubes With Screw Thread
Tubes Culture Tubes
Reusable culture tubes with screw thread made from clear borosilicate glass.
  • High resistance against chemicals
  • High thermal resistance
  • With white marking field
Ordering information: Screw caps have to be ordered separately.
Order Now
SPECIFICATIONS
Environmentally Preferable