VWR® Thermotech™, Beakers, Low Form, Heatable
Supplier: VWR Collection
VWRI213-0272EA 125 SGD
VWRI213-0272 VWRI213-0273 VWRI213-0271
VWR® Thermotech™, Beakers, Low Form, Heatable
Beakers
PTFE, white with black PTFE carbon base.
  • Totally inert pure PTFE inner surface
  • Heatable to 270 °C without distortion
  • Improved heat transfer
Features the unique combination of a pure PTFE body with a specially formulated stabilised PTFE carbon base as a single moulding.