VWR® Beakers, Low Form, PMP (TPX)
Supplier: VWR Collection
VWRI213-0510EA 13.9 SGD
VWRI213-0510 VWRI213-0482 VWRI213-0483 VWRI213-0484 VWRI213-0485 VWRI213-0486 VWRI213-0487 VWRI213-0488
VWR® Beakers, Low Form, PMP (TPX)
Beakers
Made of PMP (TPX), crystal-clear.
  • Excellent chemical resistance
  • Printed graduations
  • Withstand temperatures of 0 to 121 °C when used continuously (up to 170 °C for short periods)
  • Resin meets US FDA 21 CFR requirements
Order Now
SPECIFICATIONS
Environmentally Preferable Sustainable Packaging