VWR® Beakers, Disposable
Supplier: VWR Collection
414004-149
VWRI213-3915EA 31.6 SGD
VWRI213-3915 VWRI213-3916 VWRI213-3917 VWRI213-3918 VWRI213-3919 VWRI213-3920 VWRI213-3921 VWRI213-3922 VWRU414004-149
VWR® Beakers, Disposable
Beakers
PP, translucent.
  • Embossed graduations
  • Ribs for easy stacking
  • Autoclavable to 121 °C
  • With spout
High quality economical beaker for everyday work. Ideal for general laboratory use.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904