VWR®, PETG Erlenmeyer Shaker Flask, Baffled Bottom
Supplier: VWR Collection
VWRI214-0466EA 126 SGD
VWRI214-0466 VWRI214-0467 VWRI214-0468 VWRI214-0469 VWRI214-0470 VWRI214-0473 VWRI214-0474 VWRI214-0475
VWR®, PETG Erlenmeyer Shaker Flask, Baffled Bottom
Flasks Erlenmeyer Flasks
VWR® crystal-clear PETG Erlenmeyer shaker flasks are robust and break resistant, increasing personal safety. They are ideal for shaker culture applications and storage. The baffled bottom improves mixing and increase gas exchange.
  • Temperature range: −60 to +60 °C
  • Unlike glass, flasks will not break when dropped
  • Moulded-in graduations for accuracy
  • Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
  • Non cytotoxic, DNase-/RNase-free and non pyrogenic
  • Unique baffled design with a molded-in“1/3 Fill” line for customer
More Product Information