VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
VWRI214-1130EA 50.1 SGD
VWRI214-1130 VWRI214-1131 VWRI214-1132 VWRI214-1133 VWRI214-1134 VWRI214-1136 VWRI214-1137 VWRI214-1170 VWRI214-1171 VWRI214-1172 VWRI214-1174 VWRI214-1175 VWRI214-1173 VWRI214-0248 VWRI214-0471 VWRI214-0472
VWR®, Erlenmeyer Flasks
Flasks Erlenmeyer Flasks
Borosilicate 3.3 glass, clear.
  • Narrow or wide neck, with beaded rim
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
An economical alternative for a wide range of laboratory applications. Flasks have flared rim with markings for approximate volume graduations and a white labelling area.
ISO 1773 (narrow mouth)
DIN ISO 24450 (wide mouth)
Certifications: ISO 1773 (narrow neck), DIN ISO 24450 (wide neck).
Order Now
SPECIFICATIONS
Environmentally Preferable