VWR®, Flasks, Round Bottom, Narrow or Wide Neck
Supplier: VWR Collection
VWRI214-1182EA 18.5 SGD
VWRI214-1182 VWRI214-1177 VWRI214-1178 VWRI214-1179 VWRI214-1180 VWRI214-1181 VWRI214-1183 VWRI214-1184 VWRI214-1185 VWRI214-1186
VWR®, Flasks, Round Bottom, Narrow or Wide Neck
Flasks Round Bottom Flasks
Borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The economical alternative for a wide range of applications. Marked with capacity and a white labelling area.
ISO 1773 (narrow mouth)
DIN 12347 (wide mouth)
Certifications: ISO 1773 (narrow neck)
DIN 12347 (wide neck)
Order Now
SPECIFICATIONS
Environmentally Preferable