VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
VWRI215-1152EA 36.9 SGD
VWRI215-1152 VWRI215-1153 VWRI215-1154 VWRI215-1155 VWRI215-1156 VWRI215-1157 VWRI215-1158 VWRI215-1159 VWRI215-1160 VWRI215-1161 VWRI215-1902
VWR®, Bottles, Wide Neck, with Screw Cap
Bottles Plastic Bottles
PTFE, strong walled.
  • Very good chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Exceptionally smooth inner walls prevent contamination
Robust construction. Special recessed grips on the cap, and grooves in the flask, make it easy to handle. Non stick, the wide mouth allows easy filling and cleaning.