VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
VWRI215-1592EA 63.2 SGD
VWRI215-1592 VWRI215-1593 VWRI215-1594 VWRI215-0057 VWRI215-0058 VWRI215-0059 VWRI215-1595 VWRI215-1596 VWRI215-3261
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Bottles Glass Bottles
Borosilicate 3.3 glass, with PP screw cap and pouring ring.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Economical alternative for a wide range of laboratory applications. Round, with graduations, DIN GL 32 or GL 45 thread.
ISO 4796
Certifications: ISO 4796