VWR® Sample Vials
Supplier: VWR Collection
VWRI215-1900EA 153 SGD
VWRI215-1900 VWRI215-1901
VWR® Sample Vials
Vials
These sample vials are made from PTFE.
  • Ideal for storing aggressive or expensive materials
  • Suitable for small scale reactions at low pressure
  • Tapered inner for easy removal of contents
  • Thick wall construction
Order Now
SPECIFICATIONS
Environmentally Preferable