VWR®, Bottles, Wide Neck, HDPE, Rectangular or Square, with Screw Cap
Supplier: VWR Collection
VWRI215-1904EA 36.9 SGD
VWRI215-1904 VWRI215-1905 VWRI215-1906 VWRI215-1907 VWRI215-1908 VWRI215-1909
VWR®, Bottles, Wide Neck, HDPE, Rectangular or Square, with Screw Cap
Bottles Plastic Bottles
HDPE.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Leakproof
For collecting, storing and shipping samples, or storing liquids and dry substances. Space saving design with rectangular or square base. 2 and 4 litre bottles have integrated handle design.