VWR®, Bottle Carrier
Supplier: VWR Collection
VWRI215-3582EA 132 SGD
VWRI215-3582
VWR®, Bottle Carrier
Carriers Bottle Carriers/Containers
Glass bottles are designed to contain liquids, but when they break, hazardous liquids potentially mix with broken glass. This bottle carrier protects people and property from glass shards, whilst limiting exposure to flammable or corrosive liquids.
  • Minimises spills and breakage
  • Easy open/close mechanism
  • Environmentally friendly: Durable and liquid resistant handle construction