VWR®, Laboratory Bottles, Wide Neck
Supplier: VWR Collection
VWRI215-3743EA 21.8 SGD
VWRI215-3743 VWRI215-3744 VWRI215-3745 VWRI215-3746 VWRI215-3747 VWRI215-3748 VWRI215-3749 VWRI215-3773
VWR®, Laboratory Bottles, Wide Neck
Bottles Glass Bottles
Bottles constructed of borosilicate 3.3 glass, in clear or amber.
  • Excellent chemical resistance, high temperature resistance
  • 80 mm opening allows easy cleaning
  • Minimal thermal expansion giving relatively high resistance to temperature changes
The large opening can even accommodate large volume funnels, giving easy access to scoops or spatulas.
Amber to protect light-sensitive materials.
Certifications: In compliance with DIN ISO 4796-1 standards.
Delivery information: Supplied with screw cap and pouring ring.
Packaging: The bottles areMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable