VWR®, Dropping Bottles
Supplier: VWR Collection
VWRI215-3816EA 83.4 SGD
VWRI215-3816 VWRI215-3817 VWRI215-3818 VWRI215-3819 VWRI215-3820 VWRI215-3821 VWRI215-3822 VWRI215-3823 VWRI215-3824 VWRI215-3825 VWRI215-3826 VWRI215-3827 VWRI215-3828
VWR®, Dropping Bottles
Bottles Dropping Bottles
Soda-lime glass, amber.
  • Good chemical and physical properties
  • Pipette kit not included
Amber to protect light-sensitive substances, with DIN 18 thread.
Order Now
SPECIFICATIONS
Environmentally Preferable