VWR®, Reagent Bottles, Narrow Neck, with Standard Ground Stoppers
Supplier: VWR Collection
VWRI215-3859EA 6.9 SGD
VWRI215-3859 VWRI215-3860 VWRI215-3861 VWRI215-3862 VWRI215-3863 VWRI215-3864 VWRI215-3865 VWRI215-3866
VWR®, Reagent Bottles, Narrow Neck, with Standard Ground Stoppers
Bottles Glass Bottles
Borosilicate 3.3 glass according to ISO 4796.
  • High resistance to chemicals and thermal shock
  • Available in capacities from 10 to 2000 ml
  • Hollow glass stopper and ground joint
These reagent bottles have a precise ground neck to ensure a water- and air-tight connection between the bottle and the glass stopper. All components are made from borosilicate 3.3 glass and are, therefore, ideal for the storage of aggressive and hot media.