VWR® Aspirator Bottles, with Stopcock
Supplier: VWR Collection
VWRI215-4179EA 43 SGD
VWRI215-4179 VWRI215-4180 VWRI215-4181 VWRI215-4182
VWR® Aspirator Bottles, with Stopcock
Bottles Aspirators
Borosilicate 3.3 glass, with blue PP screw cap (GL 45) and interchangeable glass stopcock.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • All parts of the bottle are fully autoclavable
Ideal for the storage of liquids containing aggressive acids or alkalis.
Order Now
SPECIFICATIONS
Environmentally Preferable