VWR®, Wire baskets
Supplier: VWR Collection
VWRI215-4200EA 41 SGD
VWRI215-4200 VWRI215-4201 VWRI215-4202 VWRI215-4203 VWRI215-4204 VWRI215-4205 VWRI215-4206 VWRI215-4207
VWR®, Wire baskets
Baskets Wire Baskets
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Spot-welded mesh 8×8×1 mm
  • Reinforced edges
Can be used for transport, storage or sterilisation of laboratory utensils.