VWR®, Bottle Carriers
Supplier: VWR Collection
VWRI215-4208EA 77.1 SGD
VWRI215-4208 VWRI215-4209 VWRI215-4224 VWRI215-4225 VWRI215-4226 VWRI215-4227
VWR®, Bottle Carriers
Carriers Bottle Carriers/Containers
These bottle carriers are made from polyamide coated stainless steel.
  • Very good chemical and temperature resistance
  • Sturdy carrying handle
  • Four non slip feet
Suitable for safe transporting and storage of laboratory bottles.