VWR® Reagent Bottles with PP 28 Thread
Supplier: VWR Collection
VWRI215-4403EA 87.8 SGD
VWRI215-4403 VWRI215-4404 VWRI215-4405 VWRI215-4406 VWRI215-4407 VWRI215-4408 VWRI215-4409 VWRI215-4410 VWRI215-4411 VWRI215-4412 VWRI215-4413 VWRI215-4414 VWRI215-4415 VWRI215-4416 VWRI215-4417 VWRI215-4418
VWR® Reagent Bottles with PP 28 Thread
Bottles Glass Bottles
Made from clear or amber soda-lime glass. Ideal for sampling, storage, transporting liquids, or other applications that do not involve high thermal stress.
  • Good chemical and physical properties
  • Clear or amber glass
  • Amber glass bottles ideal for use with light-sensitive liquids
Bottles can be sterilised but the temperature difference (Δ T) must not exceed 40 °C.
Ordering information: Supplied without caps, screw caps must be ordered separately.
Order Now
SPECIFICATIONS
Environmentally Preferable