VWR® Reagent Bottles, Wide Neck
Supplier: VWR Collection
VWRI215-4684EA 68.9 SGD
VWRI215-4684 VWRI215-4685 VWRI215-4686 VWRI215-4687 VWRI215-4688 VWRI215-4689 VWRI215-4690 VWRI215-4691 VWRI215-4692 VWRI215-4693 VWRI215-4390 VWRI215-4391 VWRI215-4394 VWRI215-4653
VWR® Reagent Bottles, Wide Neck
Bottles Glass Bottles
Made from clear or amber soda-lime glass. Ideal for sampling, storage of powders or liquids, or other applications that do not involve high thermal stress.
  • Good chemical and physical properties
  • Clear or amber glass, delivered with black PP screw cap
  • Amber glass bottles ideal for use with light-sensitive liquids
Bottles can be sterilised but the temperature difference (Δ T) must not exceed 40 °C.
Certifications: Thread in accordance with DIN 168.
Order Now
SPECIFICATIONS
Environmentally Preferable