VWR®, Dishes, Evaporating, Tall Form
Supplier: VWR Collection
VWRI216-0179EA 41.6 SGD
VWRI216-0179 VWRI216-0180 VWRI216-0181 VWRI216-0182 VWRI216-0183 VWRI216-0184 VWRI216-0185
VWR®, Dishes, Evaporating, Tall Form
Trays/Dishes/Pots Evaporating Basins/Dishes
PTFE, with spout.
  • Very good chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Exceptionally smooth inner walls to prevent contamination
Thick walled, conical shape. Extremely robust.
Order Now
SPECIFICATIONS
Environmentally Preferable