VWR® Cooling Bricks
Supplier: VWR Collection
VWRI216-0188EA 170 SGD
VWRI216-0188 VWRI216-0189 VWRI216-0190 VWRI216-0191 VWRI216-0192 VWRI216-0193 VWRI216-0194 VWRI216-0195 VWRI216-0196
VWR® Cooling Bricks
Cooling Packs
VWR® cooling bricks are available in foam brick or gel filled pouch formats. The semi-rigid foam brick will holds its shape when refrigerated or frozen. The gel filled pouch can be refrigerated or frozen to keep products cold or frozen.
  • Long lasting, high-performance temperature protection
  • Suitable for pharmaceuticals, clinical trials, diagnostic kits, chemicals and biologicals
  • Available in a range of sizes
These packs are designed to safeguard temperature-sensitive products during storage and transport. They can be chilled in a freezer, refrigerator,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable