VWR®, Crucibles, PTFE
Supplier: VWR Collection
VWRI216-0450EA 42.1 SGD
VWRI216-0450 VWRI216-0451 VWRI216-0452 VWRI216-0453
VWR®, Crucibles, PTFE
Crucibles
Isostatically moulded in pure PTFE, FDA approved.
  • Chemically resistant and inert
  • Crack-free
  • Withstand maximum temperature of 280 °C