VWR®, Specimen Containers, Conical, with Screw Caps
Supplier: VWR Collection
VWRI216-0855EA 145 SGD
VWRI216-0855 VWRI216-0856 VWRI216-0857 VWRI216-0858 VWRI216-0859 VWRI216-0860 VWRI216-0861 VWRI216-0862 VWRI216-0863 VWRI216-0864 VWRI216-0865 VWRI216-0868 VWRI216-0869 VWRI216-1185 VWRI216-3949
VWR®, Specimen Containers, Conical, with Screw Caps
Sample Containers General Purpose Containers
PP.
  • Complete range available with different cap colours
  • Sterile as well as non sterile, individually packed or bulk
  • Frosted labelling area
General specimen containers. 120 ml capacity.
Order Now
SPECIFICATIONS
Environmentally Preferable