VWR®, Vacuum Urine Containers
Supplier: VWR Collection
VWRI216-1080EA 263 SGD
VWRI216-1080 VWRI216-1161 VWRI216-2200
VWR®, Vacuum Urine Containers
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
The VWR® urine collection container provides a faster and easier alternative for urine collection. This closed system helps prevent pouring off, leakage and wet areas, which positively impacts work environments and conditions.
  • PP, translucent
  • Frosted labelling area
  • Yellow screw cap
Urine collection container with integrated transfer device.