VWR®, Multi-Use Collection and Storage Tray
Supplier: VWR Collection
VWRI216-1492EA 184 SGD
VWRI216-1492
VWR®, Multi-Use Collection and Storage Tray
Trays/Dishes/Pots Laboratory Trays/Dishes
Plastic, blue tray with white insert.
  • Designed to hold a large number of samples, whilst being easy to carry
  • For storage on an average size refrigerator shelf
  • Hard wearing and flexible
  • Large integrated label holder for clear and easy identification
  • Can be cleaned with common disinfectants and germicides
For clinical samples. Great for transporting and storing general laboratory items. Lightweight and sturdy. Can hold up to 24 full specimen containers up to 58 mm diameter, using the insert. Removable insert to allow outer shell to be used for storageMore Product Information