VWR®, Carboys, Narrow Neck, Round, with Screw Caps
Supplier: VWR Collection
10755-110
VWRI216-1736EA 53.3 SGD
VWRI216-1736 VWRI216-1704 VWRI216-1703 VWRI216-1737 VWRI216-1706 VWRI216-1701 VWRI216-1729 VWRI216-1738 VWRI216-1702 VWRU10755-110 VWRI216-1908 VWRI216-1918 VWRI216-2180
VWR®, Carboys, Narrow Neck, Round, with Screw Caps
Bottles Carboys
LDPE, HDPE or PP, translucent with purple screw caps. Use for storing and dispensing solutions and media. Round carboys feature an integrated shoulder handle. Graduated to identify the volume dispensed.
LDPE carboys
Ideal for high purity applications as they have high resistance to chemicals (widely used in ophthalmic product range, like eye droppers and nasal sprays).
Ideal resin for cold storage (down to −80 °C), working temperature −80 to +80 °C
Resistant to physical impact (will not break even if it falls accidentally, wheras PP carboys may lead to cracking at negative temperatures)
LDPEMore Product Information