Stopcocks for VWR® Collection Carboys
Supplier: VWR Collection
VWRI216-1745EA 46.6 SGD
VWRI216-1745
Stopcocks for VWR® Collection Carboys
Stopcocks
PP, white/purple.
Pack consists of stopcock, adapter for tube connection and stopper for long storage. Stopcock is designed to be tightly assembled by hand and, therefore, does not require any tools for assembly. Autoclavable.
Resin meets US FDA 21 CFR, USP Class VI