VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWRI216-1823EA 324 SGD
VWRI216-1823 VWRI216-1808 VWRI216-1809 VWRI216-1822 VWRI216-1805 VWRI216-1806 VWRI216-1807 VWRI216-1824 VWRI216-1825 VWRI216-1826 VWRI216-1827 VWRI216-1828 VWRI216-1829 VWRI216-1830 VWRI216-1834 VWRI216-1835 VWRI216-1836 VWRI216-1837
VWR®, Straight Sample Containers, with Screw Cap
Sample Containers General Purpose Containers
PP, clear, with LDPE screw cap.
  • Excellent chemical and thermal resistance
  • Available sterile or packed under aseptic conditions
  • Four volumes are available: 40, 60, 125 and 180 ml
Straight walled containers for sample collection, storage and transport. Twist closure with leakproof inner lip for quick opening and closing. Screw caps assembled.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.