VWR®, Filling and Venting Cap for VWR® Carboys
Supplier: VWR Collection
VWRI216-1842EA 106 SGD
VWRI216-1842
VWR®, Filling and Venting Cap for VWR® Carboys
Closures Safety Caps and Safety Waste Caps
The filling and venting cap for VWR® carboys is an autoclavable closure designed for the aseptic transfer of media, biological reagents, pure water and chemicals
Each cap is equipped with integrated 6,35 mm ports.