VWR®, Carboys, Wide Neck, Round, with Screw Caps
Supplier: VWR Collection
VWRI216-1708EA 58.3 SGD
VWRI216-1708 VWRI216-1730 VWRI216-1733 VWRI216-1919
VWR®, Carboys, Wide Neck, Round, with Screw Caps
Bottles Carboys
LDPE or PP, translucent with purple screw caps.
Use for storing and dispensing solutions and media. Round carboys feature an integrated shoulder handle. Graduated to identify the volume dispensed.
LDPE carboys
Ideal for high-purity applications, as they have high resistance to chemicals (widely used in ophthalmic products like eye droppers and nasal sprays).
Preferred resin for cold storage (down to −80 °C), working temperature −80 to +80 °C
Resistant to physical impacts (will not break even if it falls down accidentally, whereas PP carboys may lead to cracking at negativeMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable