VWR®, Carboys, Round, Sanitary Neck, PP
Supplier: VWR Collection
VWRI216-1897EA 152 SGD
VWRI216-1897 VWRI216-1910 VWRI216-1920
VWR®, Carboys, Round, Sanitary Neck, PP
Bottles Carboys
Ideal for bioprocessing applications. Features a sanitary flange (76 mm) that accepts standard TC fittings.
  • Convenient shoulder handles
  • Very good chemical resistance
  • Autoclavable
Certifications: Resin meets US FDA 21 CFR, USP Class VI.
Order Now
SPECIFICATIONS
Environmentally Preferable