VWR®, Wire basket with handle
Supplier: VWR Collection
VWRI216-2254EA 202 SGD
VWRI216-2254
VWR®, Wire basket with handle
Baskets Wire Baskets
This wire basket is made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Foldaway carrying handle with plastic hand grip
  • Robust design with two rods for base reinforcement
Suitable for transportation or sterilisation of laboratory goods.