VWR® 24-Hour Urine Container
Supplier: VWR Collection
VWRI216-2327EA 179 SGD
VWRI216-2327
VWR® 24-Hour Urine Container
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
24-hour 3 L vacuum urine container with aspiration system for vacuum tubes directly integrated in the screw cap.
  • Optimal handling: Ergonomic moulded handle
  • Graduated
  • Polyethylene
The 24 hour urine collection container is suitable for collecting urine over a timed period.
The 3 L container is brown coloured to prevent the premature decomposition of light sensitive parameters such as urobilinogen.
The container is fitted with an integrated transfer device to enable hygienic, clean sample transfer into a urine tube.
Efficiency in collectingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable