VWR®, Solid Euro Containers
Supplier: VWR Collection
VWRI216-2390EA 18.4 SGD
VWRI216-2390 VWRI216-2391 VWRI216-2392 VWRI216-2393 VWRI216-2394 VWRI216-2395 VWRI216-2396 VWRI216-2397 VWRI216-2398 VWRI216-2399 VWRI216-2400 VWRI216-2401 VWRI216-2402 VWRI216-2409 VWRI216-2410 VWRI216-2416
VWR®, Solid Euro Containers
Boxes Transport Boxes
These plastic stackable Euro containers are ideal for simple storage solutions as well as highly automated systems. Made from strong plastic these containers are ideal for transporting goods and food safely. They are very stable when loaded due to reinforced frame and corner construction.
  • Made of PPC (polypropylene copolymer)
  • Suitable for every industry and for any application
  • Smooth surfaces make them easy to clean
  • Ergonomic handles for effective manual handling
  • Suitable for –20 to +65 °C storage
  • 100% recyclable
The solid Euro containers areMore Product Information