VWR® 32-Place PCR Racks
Supplier: VWR Collection
VWRI216-2988EA 140 SGD
VWRI216-2988
VWR® 32-Place PCR Racks
Racks Tube Racks
Designed for pre- and post-PCR sampling, streamline your PCR workflow with the 32-Place PCR rack. This versatile rack accommodates 0,2 ml tubes both individually and in strips of 8, as well as 32-place assay plates (8×4).
  • Small and compact design is stackable, saving valuable space in the freezer (Low temperature range: −80 °C)
  • Right sized when processing smaller batches
  • Lid hinge design allows lid to sit at 180 degrees, clear of sample processing, or be removed entirely
  • Transparent lid protects samples and allows quick assessment of contents
  • Embossed
More Product Information