VWR®, Glass Stoppers for Standard Ground Joints
Supplier: VWR Collection
VWRI217-0887EA 11.4 SGD
VWRI217-0887 VWRI217-0888 VWRI217-0889 VWRI217-0890 VWRI217-0891 VWRI217-0892 VWRI217-0893 VWRI217-0894
VWR®, Glass Stoppers for Standard Ground Joints
Stoppers Glass Stoppers
Borosilicate 3.3 glass.
  • Hollow, with hexagonal grip
  • Trip-tip to ensure every last drop of liquid returns into the vessel
  • Resistant to chemicals and thermal shock
Certifications: DIN 12252
Order Now
SPECIFICATIONS
Environmentally Preferable