VWR®, Funnels with Short Stem
Supplier: VWR Collection
VWRI221-0175EA 4.1 SGD
VWRI221-0175 VWRI221-0176 VWRI221-0177 VWRI221-0178 VWRI221-0179 VWRI221-0180 VWRI221-0181 VWRI221-0182
VWR®, Funnels with Short Stem
Funnels Filtration Funnels Filter Funnels
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Smooth internal walls.
Certifications: ISO 4798.
Order Now
SPECIFICATIONS
Environmentally Preferable