VWR®, Funnels, Powder
Supplier: VWR Collection
414004-272
VWRU414004-272EA 42 SGD
VWRU414004-272 VWRI221-0239 VWRI221-0240 VWRI221-0241 VWRI221-0242
VWR®, Funnels, Powder
Funnels Powder Funnels
PP, translucent.
  • Parallel stem minimises powder bridging
  • External ribs for better air evacuation
  • Excellent chemical and temperature resistance
  • Autoclavable
Used for transferring powders. Also suitable for liquids.
Order Now
SPECIFICATIONS
Environmentally Preferable