VWR®, Disposable Paper Funnel
Supplier: VWR Collection
VWRI221-0274EA 66.5 SGD
VWRI221-0274
VWR®, Disposable Paper Funnel
Funnels Filtration Funnels Filter Funnels
Made of clay-coated heavy paper stock.
  • Innovative 'backrest' allows hands-free use when pouring viscous liquids
  • Fits a wide variety of openings
  • Maximum flexibility - Cut stem to any opening size or shape
  • Apply a minimum of pressure and the funnel will stay in position
  • Use with virtually any liquid or powder*
Good alternative to disposable plastic funnels. Environmentally friendly paper product will naturally biodegrade in the standard municipal waste stream. Helps to reduce cross contamination.
Caution: * Not recommended for hot liquids.