VWR®, Scoop
Supplier: VWR Collection
VWRI231-0501EA 66.1 SGD
VWRI231-0501
VWR®, Scoop
Scoops
PTFE, white.
  • Totally chemically inert
  • Non contaminating
  • Suitable for use with frozen substances and other solids
Scoop length: 80 mm
Total length: 140 mm
Order Now
SPECIFICATIONS
Environmentally Preferable