VWR®, Laboratory Jacks
Supplier: VWR Collection
VWRI245-0004EA 444 SGD
VWRI245-0004 VWRI245-0005 VWRI245-0006 VWRI245-0007 VWRI245-0008 VWRI245-0009
VWR®, Laboratory Jacks
Laboratory Jacks
Anodised aluminium. With M10 threaded hole in the middle of the top plate for attaching retort rods (starting from item number 245-0006).
  • Smooth adjustment of working height
  • Extreme stability
  • Exceptionally sturdy
  • Corrosion resistant and non susceptible to a wide range of chemicals